missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol/Ether Blend, 50% v/v, Reagents
Methanol/Ether Blend, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$463.43 - $927.73
Chemical Identifiers
| CAS | 67-56-1 |
|---|---|
| Molecular Formula | CH4O |
| Molecular Weight (g/mol) | 32.04 |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| IUPAC Name | methanol |
| SMILES | CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-334-776
|
Labchem Inc
CM0104001C |
1 L |
Each for $463.43
|
|
|||||
|
01-334-775
|
Labchem Inc
CM0104004C |
4 L |
Each for $927.73
|
|
|||||
Specifications
| 67-56-1,60-29-7 | |
| 35°C to 64.7°C | |
| Glass Bottle | |
| 0.7524 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 50% | |
| Methanol/Ether Blend, 50% v/v |
| Colorless | |
| 1 L | |
| Liquid | |
| CH4O | |
| CO | |
| 32.04 | |
| Reagent |