missing translation for 'onlineSavingsMsg'
Learn More

Methanol/Ether Blend, 50% v/v, Reagents

Methanol/Ether Blend, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc CM0104001C

 View more versions of this product

Catalog No. 01-334-776


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methanol/Ether Blend, 50% v/v
Colorless
1 L
Liquid
CH4O
CO
32.04
Reagent
67-56-1,60-29-7
35°C to 64.7°C
Glass Bottle
0.7524
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
50%

Chemical Identifiers

67-56-1
32.04
methanol
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
CO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions