missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol/Ether Blend, 50% v/v, Reagents
Methanol/Ether Blend, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CM0104004C
Specifications
| Methanol/Ether Blend, 50% v/v | |
| Colorless | |
| 4 L | |
| Liquid | |
| CH4O | |
| CO | |
| 32.04 | |
| Reagent |
| 67-56-1,60-29-7 | |
| 35°C to 64.7°C | |
| Glass Bottle | |
| 0.7524 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 50% |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| methanol |
| CH4O | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| CO |