missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic Acid, Technical, 69.5-70.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M2017200LTBL

 View more versions of this product

Catalog No. 18611651

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

75-75-2
Drum
200 L
CS(O)(=O)=O
96.10
Technical
1
CH4O3S
AFVFQIVMOAPDHO-UHFFFAOYSA-N
methanesulfonic acid
69.5 to 70.5%

Chemical Identifiers

75-75-2
96.10
methanesulfonic acid
CH4O3S
AFVFQIVMOAPDHO-UHFFFAOYSA-N
CS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.