missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic Acid, Technical, 69.5-70.5%, Spectrum™ Chemical

Catalog No. p-7165894
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611654 500 mL Amber Glass Bottle
18611652 4 L Amber Glass Bottle
18611650 20 L Poly Drum
18611651 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611654 Supplier Spectrum Chemical Mfg Cor Supplier No. M2017500MLGL

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.10
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
IUPAC Name methanesulfonic acid
SMILES CS(O)(=O)=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 69.5 to 70.5%
Grade Technical

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.