missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methacholine Chloride 98.0+%, TCI America™
$171.34 - $1638.33
Chemical Identifiers
| CAS | 62-51-1 |
|---|---|
| Molecular Formula | C8H18ClNO2 |
| Molecular Weight (g/mol) | 195.687 |
| MDL Number | MFCD00011817 |
| InChI Key | JHPHVAVFUYTVCL-UHFFFAOYSA-M |
| Synonym | methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride |
| PubChem CID | 6114 |
| ChEBI | CHEBI:50142 |
| IUPAC Name | 2-acetyloxypropyl(trimethyl)azanium;chloride |
| SMILES | CC(C[N+](C)(C)C)OC(=O)C.[Cl-] |
Chemical Identifiers
| 62-51-1 | |
| 195.687 | |
| JHPHVAVFUYTVCL-UHFFFAOYSA-M | |
| 6114 | |
| 2-acetyloxypropyl(trimethyl)azanium;chloride |
| C8H18ClNO2 | |
| MFCD00011817 | |
| methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride | |
| CHEBI:50142 | |
| CC(C[N+](C)(C)C)OC(=O)C.[Cl-] |
Specifications
| 62-51-1 | |
| White | |
| MFCD00011817 | |
| methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride | |
| CC(C[N+](C)(C)C)OC(=O)C.[Cl-] | |
| 195.687 | |
| CHEBI:50142 | |
| ≥98.0% (T) | |
| Methacholine Chloride |
| 172°C | |
| C8H18ClNO2 | |
| 25 g | |
| JHPHVAVFUYTVCL-UHFFFAOYSA-M | |
| 2-acetyloxypropyl(trimethyl)azanium;chloride | |
| 6114 | |
| 195.69 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1402
RTECSNumber : BR5250000
TSCA : Yes