missing translation for 'onlineSavingsMsg'
Learn More

Methacholine Chloride 98.0+%, TCI America™

Supplier:  TCI America M007325G

 View more versions of this product

Catalog No. M007325G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methacholine Chloride
172°C
C8H18ClNO2
25 g
JHPHVAVFUYTVCL-UHFFFAOYSA-M
2-acetyloxypropyl(trimethyl)azanium;chloride
6114
195.69
Crystalline Powder
62-51-1
White
MFCD00011817
methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride
CC(C[N+](C)(C)C)OC(=O)C.[Cl-]
195.687
CHEBI:50142
≥98.0% (T)

Chemical Identifiers

62-51-1
195.687
JHPHVAVFUYTVCL-UHFFFAOYSA-M
6114
2-acetyloxypropyl(trimethyl)azanium;chloride
C8H18ClNO2
MFCD00011817
methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride
CHEBI:50142
CC(C[N+](C)(C)C)OC(=O)C.[Cl-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1402

RTECSNumber : BR5250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions