missing translation for 'onlineSavingsMsg'
Learn More

Metaphosphoric Acid, 3% w/v, Reagents

Catalog No. 01334760
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334760 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334760 Supplier Labchem Inc Supplier No. CM103500500A
Only null left

Metaphosphoric Acid, 3% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Phosphenic Acid
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.019
Quantity 500 mL
Physical Form Liquid
Percent Purity 3%
Grade Reagent
Chemical Name or Material Metaphosphoric Acid, 3% w/v