missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Metamizole sodium monohydrate

Catalog No. AC467482500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467482500 250 mg
AC467480010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467482500 Supplier Thermo Scientific Chemicals Supplier No. 467482500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5907-38-0
Molecular Formula C13H16N3NaO4S·H2O

Specifications

Quantity 250 mg
Solubility Solubility aq. soln.: soluble. Other solubilities: soluble in methanol, insoluble in ether, acetone, chloroform
Formula Weight 351.36
Chemical Name or Material Metamizole sodium monohydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Suspected of damaging fertility or the unborn child.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only