missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesotartaric Acid Monohydrate 90.0+%, TCI America™
$303.84 - $965.59
Chemical Identifiers
| CAS | 5990-63-6 |
|---|---|
| Molecular Formula | C4H8O7 |
| Molecular Weight (g/mol) | 168.101 |
| MDL Number | MFCD00063641 |
| InChI Key | UUDLQDCYDSATCH-NUGIMEKKSA-N |
| Synonym | meso-Tartaric Acid |
| PubChem CID | 16218804 |
| IUPAC Name | (2R,3S)-2,3-dihydroxybutanedioic acid;hydrate |
| SMILES | C(C(C(=O)O)O)(C(=O)O)O.O |
Chemical Identifiers
| 5990-63-6 | |
| 168.101 | |
| UUDLQDCYDSATCH-NUGIMEKKSA-N | |
| 16218804 | |
| C(C(C(=O)O)O)(C(=O)O)O.O |
| C4H8O7 | |
| MFCD00063641 | |
| meso-Tartaric Acid | |
| (2R,3S)-2,3-dihydroxybutanedioic acid;hydrate |
Specifications
| 5990-63-6 | |
| White | |
| MFCD00063641 | |
| meso-Tartaric Acid | |
| C(C(C(=O)O)O)(C(=O)O)O.O | |
| 168.101 | |
| 150.09 | |
| Crystalline Powder |
| 75°C | |
| C4H8O7 | |
| 5 g | |
| UUDLQDCYDSATCH-NUGIMEKKSA-N | |
| (2R,3S)-2,3-dihydroxybutanedioic acid;hydrate | |
| 16218804 | |
| ≥90.0% (HPLC) | |
| Mesotartaric Acid Monohydrate |
Safety and Handling
EINECSNumber : (2)-1456
TSCA : Yes