missing translation for 'onlineSavingsMsg'
Learn More

Mesotartaric Acid Monohydrate 90.0+%, TCI America™

Supplier:  TCI America M007025G

 View more versions of this product

Catalog No. M007025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Mesotartaric Acid Monohydrate
75°C
C4H8O7
25 g
UUDLQDCYDSATCH-NUGIMEKKSA-N
(2R,3S)-2,3-dihydroxybutanedioic acid;hydrate
16218804
≥90.0% (HPLC)
5990-63-6
White
MFCD00063641
meso-Tartaric Acid
C(C(C(=O)O)O)(C(=O)O)O.O
168.101
150.09
Crystalline Powder

Chemical Identifiers

5990-63-6
168.101
UUDLQDCYDSATCH-NUGIMEKKSA-N
16218804
C(C(C(=O)O)O)(C(=O)O)O.O
C4H8O7
MFCD00063641
meso-Tartaric Acid
(2R,3S)-2,3-dihydroxybutanedioic acid;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1456

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions