accessibility menu, dialog, popup

UserName

OOHOHOOHOHH2O

CAS RN 5990-63-6

IUPAC Name: 2,3-dihydroxybutanedioic acid hydrate
Synonyms: (.+-.)-tartaric acid hydrate
Molecular Weight (g/mol): 168.1
Molecular Formula: C4H8O7
InChi Key: UUDLQDCYDSATCH-UHFFFAOYNA-N
SMILES: O.OC(C(O)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results