missing translation for 'onlineSavingsMsg'
Learn More
Learn More
meso-Erythritol 99.0+%, TCI America™
$70.25 - $613.25
Chemical Identifiers
| CAS | 149-32-6 |
|---|---|
| Molecular Formula | C4H10O4 |
| Molecular Weight (g/mol) | 122.12 |
| MDL Number | MFCD00004710 |
| InChI Key | UNXHWFMMPAWVPI-UHFFFAOYNA-N |
| Synonym | erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite |
| PubChem CID | 222285 |
| ChEBI | CHEBI:17113 |
| IUPAC Name | butane-1,2,3,4-tetrol |
| SMILES | OCC(O)C(O)CO |
Chemical Identifiers
| 149-32-6 | |
| 122.12 | |
| UNXHWFMMPAWVPI-UHFFFAOYNA-N | |
| 222285 | |
| butane-1,2,3,4-tetrol |
| C4H10O4 | |
| MFCD00004710 | |
| erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite | |
| CHEBI:17113 | |
| OCC(O)C(O)CO |
Specifications
| 149-32-6 | |
| White | |
| C4H10O4 | |
| 25 g | |
| UNXHWFMMPAWVPI-UHFFFAOYNA-N | |
| butane-1,2,3,4-tetrol | |
| 222285 | |
| 122.12 | |
| Crystalline Powder |
| 120°C | |
| 331°C | |
| MFCD00004710 | |
| erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite | |
| OCC(O)C(O)CO | |
| 122.12 | |
| CHEBI:17113 | |
| ≥99.0% (HPLC,T) | |
| meso-Erythritol |
Safety and Handling
EINECSNumber : (2)-3651
RTECSNumber : KF2000000
TSCA : Yes