missing translation for 'onlineSavingsMsg'
Learn More

meso-Erythritol 99.0+%, TCI America™

Supplier:  TCI America E0021500G

 View more versions of this product

Catalog No. E0021500G


Only null left

Description

Description

Specifications

Specifications

meso-Erythritol
120°C
331°C
MFCD00004710
erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite
OCC(O)C(O)CO
122.12
CHEBI:17113
≥99.0% (HPLC,T)
149-32-6
White
C4H10O4
500 g
UNXHWFMMPAWVPI-UHFFFAOYNA-N
butane-1,2,3,4-tetrol
222285
122.12
Crystalline Powder

Chemical Identifiers

149-32-6
122.12
UNXHWFMMPAWVPI-UHFFFAOYNA-N
222285
butane-1,2,3,4-tetrol
C4H10O4
MFCD00004710
erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite
CHEBI:17113
OCC(O)C(O)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3651

RTECSNumber : KF2000000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions