missing translation for 'onlineSavingsMsg'
Learn More
Learn More
meso-1,2,3,4-Tetrabromobutane 98.0+%, TCI America™
$80.85 - $635.31
Chemical Identifiers
| CAS | 2657-67-2 |
|---|---|
| Molecular Formula | C4H6Br4 |
| Molecular Weight (g/mol) | 373.708 |
| MDL Number | MFCD00070471 |
| InChI Key | HGRZLIGHKHRTRE-ZXZARUISSA-N |
| PubChem CID | 22820355 |
| IUPAC Name | (2S,3R)-1,2,3,4-tetrabromobutane |
| SMILES | C(C(C(CBr)Br)Br)Br |
Chemical Identifiers
| 2657-67-2 | |
| 373.708 | |
| HGRZLIGHKHRTRE-ZXZARUISSA-N | |
| (2S,3R)-1,2,3,4-tetrabromobutane |
| C4H6Br4 | |
| MFCD00070471 | |
| 22820355 | |
| C(C(C(CBr)Br)Br)Br |
Specifications
| 2657-67-2 | |
| White | |
| C4H6Br4 | |
| 25 g | |
| C(C(C(CBr)Br)Br)Br | |
| 373.708 | |
| 373.71 | |
| Crystalline Powder |
| 116°C | |
| 181°C | |
| MFCD00070471 | |
| HGRZLIGHKHRTRE-ZXZARUISSA-N | |
| (2S,3R)-1,2,3,4-tetrabromobutane | |
| 22820355 | |
| ≥98.0% (GC) | |
| meso-1,2,3,4-Tetrabromobutane |
Safety and Handling
EINECSNumber : (2)-2364
TSCA : No