accessibility menu, dialog, popup

UserName

BrBrBrBr

CAS RN 2657-67-2

IUPAC Name: 1,2,3,4-tetrabromobutane
Synonyms: 1,2,3,4-tetrabromobutane
Molecular Weight (g/mol): 373.71
Molecular Formula: C4H6Br4
InChi Key: HGRZLIGHKHRTRE-UHFFFAOYNA-N
SMILES: BrCC(Br)C(Br)CBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results