missing translation for 'onlineSavingsMsg'
Learn More

Mesityl Iodide 98.0+%, TCI America™

Supplier:  TCI America I04785G

 View more versions of this product

Catalog No. I04785G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Mesityl Iodide
31°C
250°C
MFCD00013707
2,4,6-trimethyliodobenzene, 2-iodomesitylene, mesityl iodide, benzene, 2-iodo-1,3,5-trimethyl, iodo-2,4,6-trimethylbenzene, 2,4,6-trimethyliodobenzene 2-iodomesitylene, iodomesitylene, jodmesitylen, mesityliodine, pubchem12601
CC1=CC(=C(C(=C1)C)I)C
246.091
246.09
Crystalline Powder
4028-63-1
White
C9H11I
5 g
GTPNXFKONRIHRW-UHFFFAOYSA-N
2-iodo-1,3,5-trimethylbenzene
77647
≥98.0% (GC)

Chemical Identifiers

4028-63-1
246.091
GTPNXFKONRIHRW-UHFFFAOYSA-N
77647
CC1=CC(=C(C(=C1)C)I)C
C9H11I
MFCD00013707
2,4,6-trimethyliodobenzene, 2-iodomesitylene, mesityl iodide, benzene, 2-iodo-1,3,5-trimethyl, iodo-2,4,6-trimethylbenzene, 2,4,6-trimethyliodobenzene 2-iodomesitylene, iodomesitylene, jodmesitylen, mesityliodine, pubchem12601
2-iodo-1,3,5-trimethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions