missing translation for 'onlineSavingsMsg'
Learn More

Mesityl Iodide 98.0+%, TCI America™

Catalog No. p-7131431
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04785G 5 g
I047825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04785G Supplier TCI America Supplier No. I04785G

Chemical Identifiers

CAS 4028-63-1
Molecular Formula C9H11I
Molecular Weight (g/mol) 246.091
MDL Number MFCD00013707
InChI Key GTPNXFKONRIHRW-UHFFFAOYSA-N
Synonym 2,4,6-trimethyliodobenzene, 2-iodomesitylene, mesityl iodide, benzene, 2-iodo-1,3,5-trimethyl, iodo-2,4,6-trimethylbenzene, 2,4,6-trimethyliodobenzene 2-iodomesitylene, iodomesitylene, jodmesitylen, mesityliodine, pubchem12601
PubChem CID 77647
IUPAC Name 2-iodo-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)I)C

Specifications

Melting Point 31°C
Color White
Boiling Point 250°C
Quantity 5 g
Formula Weight 246.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Mesityl Iodide
TSCA No
Recommended Storage Refrigerator