missing translation for 'onlineSavingsMsg'
Learn More

Mesityl Fluoride 98.0+%, TCI America™

Catalog No. M12011G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12011G 1 g
M12015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12011G Supplier TCI America Supplier No. M12011G

Chemical Identifiers

CAS 392-69-8
Molecular Formula C9H11F
Molecular Weight (g/mol) 138.19
MDL Number MFCD00052365
InChI Key ZLGPNBBJPOBSLY-UHFFFAOYSA-N
Synonym fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939
PubChem CID 136234
IUPAC Name 2-fluoro-1,3,5-trimethylbenzene
SMILES CC1=CC(C)=C(F)C(C)=C1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 1 g
Formula Weight 138.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Mesityl Fluoride