missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesityl Bromide 99.0+%, TCI America™
Supplier: TCI America B1261100G
Specifications
| Mesityl Bromide | |
| Yellow | |
| C9H11Br | |
| 100 g | |
| RRTLQRYOJOSPEA-UHFFFAOYSA-N | |
| 2-bromo-1,3,5-trimethylbenzene | |
| 68473 | |
| ≥99.0% (GC) |
| 576-83-0 | |
| 225°C | |
| MFCD00000073 | |
| 2-bromomesitylene, 2,4,6-trimethylbromobenzene, mesityl bromide, bromomesitylene, benzene, 2-bromo-1,3,5-trimethyl, 2,4,6-trimethybromombenzene, mesitylene, 2-bromo, 2,4,6-trimethylphenyl bromide, 1-bromo-2,4,6-trimethylbenzene, 2-bromo-1,3,5-trimethyl-benzene | |
| CC1=CC(=C(C(=C1)C)Br)C | |
| 199.091 | |
| 199.09 | |
| Liquid |
Chemical Identifiers
| 576-83-0 | |
| 199.091 | |
| RRTLQRYOJOSPEA-UHFFFAOYSA-N | |
| 68473 | |
| CC1=CC(=C(C(=C1)C)Br)C |
| C9H11Br | |
| MFCD00000073 | |
| 2-bromomesitylene, 2,4,6-trimethylbromobenzene, mesityl bromide, bromomesitylene, benzene, 2-bromo-1,3,5-trimethyl, 2,4,6-trimethybromombenzene, mesitylene, 2-bromo, 2,4,6-trimethylphenyl bromide, 1-bromo-2,4,6-trimethylbenzene, 2-bromo-1,3,5-trimethyl-benzene | |
| 2-bromo-1,3,5-trimethylbenzene |
Safety and Handling
EINECSNumber : (3)-0057
TSCA : Yes