Please login to your online account to display your discounted pricing

MES, ULTROL™ Grade, Calbiochem™, EMD Millipore

A zwitterionic buffer useful in the pH range of 5.5 to 6.5

$86.00 - $630.00

Chemical Identifiers

Assay Percent Range ≥99.0%
Formula Weight 195.2
Buffers zwitterionic buffer
CAS 4432-31-9
Chemical Name or Material MES
Molecular Formula C6H13NO4S
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

475893100GM

 
emd millipore corporation
475893100GM
Each for $86.00

4758931KG

 
emd millipore corporation
4758931KG
Each for $630.00

475893500GM

 
emd millipore corporation
475893500GM
Each for $413.00
Description & Specifications

Specifications

Formula Weight 195.2
Quantity 100g
Chemical Name or Material MES
Buffers zwitterionic buffer
Purity Grade Notes by titration (dry basis)
Assay Percent Range ≥99.0%
Molecular Formula C6H13NO4S
CAS 4432-31-9
Synonym 2-(N-Morpholino)ethanesulfonic Acid
Solubility Information Soluble in water
Purity ULTROL™ Grade
Recommended Storage 15°C to 30°C (Do not freeze).
Safety and Handling

15°C to 30°C (Do not freeze).