missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MES, Sodium Salt, ULTROL™ Grade, Calbiochem™,
A zwitterionic buffer useful in the pH range of 5.5 to 6.7
$1,320.00 - $1,320.00
Chemical Identifiers
| CAS | 71119-23-8 |
|---|---|
| Molecular Formula | C6H12NNaO4S |
| Molecular Weight (g/mol) | 217.215 |
| InChI Key | IRHWMYKYLWNHTL-UHFFFAOYSA-M |
| Synonym | 2-(N-Morpholino)ethanesulfonic Acid, Na |
| PubChem CID | 23673676 |
| ChEBI | CHEBI:62955 |
| IUPAC Name | sodium;2-morpholin-4-ylethanesulfonate |
| SMILES | C1COCCN1CCS(=O)(=O)[O-].[Na+] |
Chemical Identifiers
| 71119-23-8 | |
| 217.215 | |
| 2-(N-Morpholino)ethanesulfonic Acid, Na | |
| CHEBI:62955 | |
| C1COCCN1CCS(=O)(=O)[O-].[Na+] |
| C6H12NNaO4S | |
| IRHWMYKYLWNHTL-UHFFFAOYSA-M | |
| 23673676 | |
| sodium;2-morpholin-4-ylethanesulfonate |
Specifications
| 71119-23-8 | |
| ≥99.0% | |
| 2-(N-Morpholino)ethanesulfonic Acid, Na | |
| IRHWMYKYLWNHTL-UHFFFAOYSA-M | |
| sodium;2-morpholin-4-ylethanesulfonate | |
| 23673676 | |
| 217.2 | |
| zwitterionic buffer |
| 1 kg | |
| C6H12NNaO4S | |
| Soluble in water | |
| C1COCCN1CCS(=O)(=O)[O-].[Na+] | |
| 217.215 | |
| CHEBI:62955 | |
| by titration (dry basis) |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze).