missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MES, Sodium Salt, ULTROL™ Grade, Calbiochem™,

Catalog No. 4758941KG
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4758941KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4758941KG Supplier MilliporeSigma™ Supplier No. 4758941KG
Only null left

A zwitterionic buffer useful in the pH range of 5.5 to 6.7

Chemical Identifiers

CAS 71119-23-8
Molecular Formula C6H12NNaO4S
Molecular Weight (g/mol) 217.215
InChI Key IRHWMYKYLWNHTL-UHFFFAOYSA-M
Synonym 2-(N-Morpholino)ethanesulfonic Acid, Na
PubChem CID 23673676
ChEBI CHEBI:62955
IUPAC Name sodium;2-morpholin-4-ylethanesulfonate
SMILES C1COCCN1CCS(=O)(=O)[O-].[Na+]

Specifications

Quantity 1 kg
Assay Percent Range ≥99.0%
Solubility Information Soluble in water
Formula Weight 217.2
Grade by titration (dry basis)
Chemical Name or Material zwitterionic buffer
Recommended Storage 15°C to 30°C (Do not freeze).