missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals MES sodium salt, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
| Absorbance | 0.03 max. (10% in water) 1 cm cell vs water at 280nm, 0.05 max. (10% in water) 1 cm cell vs water at 260nm |
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | 99% |
| Infrared Spectrum | Authentic |
| Packaging | Plastic bottle |
| Solubility Information | Solubility in water: 0.5g/L. |
| Formula Weight | 217.22 |
| Percent Purity | ≥98.5% (after ion exchange) |
| Show More |
RUO – Research Use Only