missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES hydrate, 99+%, for biochemistry

CAS: 1266615-59-1 | C6H13NO4S | 195.23 g/mol

$121.63 - $3400.62

Chemical Identifiers

CAS 1266615-59-1
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.23
MDL Number MFCD00149409
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 4-Morpholinoethanesulfonic acid hydrate, MES
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid
SMILES OS(=O)(=O)CCN1CCOCC1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC172590250
SDS View Documents
Thermo Scientific Chemicals
172590250
25 g Plastic bottle
Each for $121.63
Only null left
 
AC172591000
SDS View Documents
Thermo Scientific Chemicals
172591000
100 g Plastic bottle
Each for $280.53
Only null left
 
AC172595000
SDS View Documents
Thermo Scientific Chemicals
172595000
500 g Plastic bottle
Each for $1,076.73
Only null left
 
AC172590025
SDS View Documents
Thermo Scientific Chemicals
172590025
2.5 kg Plastic bottle
Each for $3,400.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1266615-59-1
195.23
SXGZJKUKBWWHRA-UHFFFAOYSA-N
2-(morpholin-4-yl)ethane-1-sulfonic acid
C6H13NO4S
MFCD00149409
4-Morpholinoethanesulfonic acid hydrate, MES
OS(=O)(=O)CCN1CCOCC1

Specifications

(0.1M in H2O) 1 cm cell, (0.1M in H2O) 1 cm cell at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
243.0°C
White
Needle-like Crystals or Powder
99+%
9.5% max. (105°C, 3 hrs)
MFCD00149409
14, 5902
Solubility in water: soluble.
OS(=O)(=O)CCN1CCOCC1
195.23
≥99%
MES hydrate, 0.99
(0.1M aq. soln.) (1 cm cell)
1266615-59-1
2.5 to 4.0 (1%, H2O, 25°C)
25 g
Authentic
C6H13NO4S
Plastic bottle
4-Morpholinoethanesulfonic acid hydrate, MES
SXGZJKUKBWWHRA-UHFFFAOYSA-N
2-(morpholin-4-yl)ethane-1-sulfonic acid
195.23
Biochemical
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

EINECSNumber : 224-632-3

RTECSNumber : QE3479500

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only