missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals MES hydrate, 99+%, for biochemistry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Absorbance | (0.1M in H2O) 1 cm cell, (0.1M in H2O) 1 cm cell at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm |
| Absorbance Conditions | (0.1M aq. soln.) (1 cm cell) |
| Melting Point | 243.0°C |
| Color | White |
| pH | 2.5 to 4.0 (1%, H2O, 25°C) |
| Physical Form | Needle-like Crystals or Powder |
| Quantity | 500 g |
| Assay Percent Range | 99+% |
| Infrared Spectrum | Authentic |
| Loss on Drying | 9.5% max. (105°C, 3 hrs) |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| EINECSNumber | 224-632-3 |
| RTECSNumber | QE3479500 |
| TSCA | TSCA |
RUO – Research Use Only