missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ MES, free acid
Description
MES, free acid is a commonly used as a buffer.
Specifications
Specifications
| Chemical Name or Material | 2-(N-Morpholino)ethanesulfonic acid |
| CAS | 4432-31-9 |
| Assay Percent Range | >99% |
| Quantity | 250 g |
| Product Line | Tocris |
| Molecular Formula | C6H13NO4S |
| Formula Weight | Observed MW: 195.24 |