missing translation for 'onlineSavingsMsg'
Learn More

Mercury Standard For ICP/MS, 10ppm in 5% HNO3, SPEX CertiPrep™

Catalog No. CLHG21AM
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLHG21AM 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLHG21AM Supplier SPEX CertiPrep Group LLC Supplier No. CLHG21AM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Hg10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Solvent or Matrix 5% HNO3
Quantity 30 mL
Chemical Name or Material Mercury Standard
Hazard Category Causes skin irritation, serious eye irritation
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only