missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury Standard For ICP, 1000μg/mL in 10% HNO3, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLHG42M
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Hg | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Mercury Standard | |
| 7697-37-2 | |
| HNO3 | |
| AA, ICP | |
| Certified Reference Material | |
| O[N+]([O-])=O | |
| nitric acid | |
| 30 mL | |
| CHEBI:48107 |
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| MFCD00011349 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 10% HNO3 | |
| 63.01 | |
| 944 |
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
| HNO3 | |
| MFCD00011349 | |
| 944 | |
| nitric acid |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only