missing translation for 'onlineSavingsMsg'
Learn More

Mercuric Acetate TS, 6% (w/v) in Glacial Acetic Acid, Ricca Chemical

Catalog No. 462932
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
462932 1 L Amber Glass Bottle
462916 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. 462932 Supplier Ricca Chemical Company Supplier No. 462932

missing translation for 'validMainframeMsg'

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 1600-27-7 , 64-19-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Color Colorless
CAS Min % 93.0, 5.3
CAS Max % 96.0, 5.52
Percent Purity 6%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Glacial Acetic Acid
pH <2
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 6% (w/v)
Chemical Name or Material Mercuric Acetate, Test Solution
Show More Show Less