missing translation for 'onlineSavingsMsg'
Learn More

Mercuric Acetate, 25 g/L in Glacial Acetic Acid, Ricca Chemical

Catalog No. p-3251907
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Safety-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R46250001C1 1 L Safety-coated Glass Bottle
46251 4 L Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. R46250001C1 Supplier Ricca Chemical Company Supplier No. R46250001C1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 1600-27-7 , 64-19-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Color Colorless
CAS Min % 96.0, 2.24
CAS Max % 100.0, 2.33
Grade Laboratory
Packaging Safety-coated Glass Bottle
Solvent or Matrix Glacial Acetic Acid
pH <2
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 25g/L
Chemical Name or Material Mercuric Acetate
Show More Show Less