missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mephenytoin

Catalog No. AC466690250
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466690250 25 mg
AC466691000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466690250 Supplier Thermo Scientific Chemicals Supplier No. 466690250

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-12-4
Molecular Formula C12H14N2O2

Specifications

Quantity 25 mg
Solubility Solubility aq. soln.: insoluble.
Melting Point 135.0°C to 137.0°C
Formula Weight 218.25
InChI Key GMHKMTDVRCWUDX-UHFFFAOYNA-N
SMILES CCC1(NC(=O)N(C)C1=O)C1=CC=CC=C1
IUPAC Name 5-ethyl-3-methyl-5-phenylimidazolidine-2,4-dione
Molecular Weight (g/mol) 218.26
Chemical Name or Material Mephenytoin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 200-012-8

RUO – Research Use Only