missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Menthyl Acetate 98.0+%, TCI America™
$98.18 - $338.84
Chemical Identifiers
| CAS | 5157-89-1 |
|---|---|
| Molecular Formula | C12H22O2 |
| Molecular Weight (g/mol) | 198.306 |
| MDL Number | MFCD00062977 |
| InChI Key | XHXUANMFYXWVNG-DLOVCJGASA-N |
| Synonym | neoisomenthyl acetate, +-menthylacetate, 1s-2alpha-1-methylethyl-5alpha-methylcyclohexan-1alpha-ol acetate |
| PubChem CID | 21630934 |
| IUPAC Name | [(1S,2S,5S)-5-methyl-2-propan-2-ylcyclohexyl] acetate |
| SMILES | CC1CCC(C(C1)OC(=O)C)C(C)C |
Chemical Identifiers
| 5157-89-1 | |
| 198.306 | |
| XHXUANMFYXWVNG-DLOVCJGASA-N | |
| 21630934 | |
| CC1CCC(C(C1)OC(=O)C)C(C)C |
| C12H22O2 | |
| MFCD00062977 | |
| neoisomenthyl acetate, +-menthylacetate, 1s-2alpha-1-methylethyl-5alpha-methylcyclohexan-1alpha-ol acetate | |
| [(1S,2S,5S)-5-methyl-2-propan-2-ylcyclohexyl] acetate |
Specifications
| 5157-89-1 | |
| 229°C | |
| MFCD00062977 | |
| neoisomenthyl acetate, +-menthylacetate, 1s-2alpha-1-methylethyl-5alpha-methylcyclohexan-1alpha-ol acetate | |
| CC1CCC(C(C1)OC(=O)C)C(C)C | |
| 198.306 | |
| 198.31 | |
| Liquid |
| Colorless | |
| C12H22O2 | |
| 1 mL | |
| XHXUANMFYXWVNG-DLOVCJGASA-N | |
| [(1S,2S,5S)-5-methyl-2-propan-2-ylcyclohexyl] acetate | |
| 21630934 | |
| ≥98.0% (GC) | |
| (+)-Menthyl Acetate |
Safety and Handling
TSCA : No