accessibility menu, dialog, popup

UserName

CAS RN 5157-89-1

IUPAC Name: 5-methyl-2-(propan-2-yl)cyclohexyl acetate
Synonyms: (-)-menthyl acetate dl-MENTHYL ACETATE
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: XHXUANMFYXWVNG-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1OC(C)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)