missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Menthyl Acetate 98.0+%, TCI America™
Supplier: TCI America M26315ML
Specifications
| (+)-Menthyl Acetate | |
| Colorless | |
| C12H22O2 | |
| 5 mL | |
| XHXUANMFYXWVNG-DLOVCJGASA-N | |
| [(1S,2S,5S)-5-methyl-2-propan-2-ylcyclohexyl] acetate | |
| 21630934 | |
| ≥98.0% (GC) |
| 5157-89-1 | |
| 229°C | |
| MFCD00062977 | |
| neoisomenthyl acetate, +-menthylacetate, 1s-2alpha-1-methylethyl-5alpha-methylcyclohexan-1alpha-ol acetate | |
| CC1CCC(C(C1)OC(=O)C)C(C)C | |
| 198.306 | |
| 198.31 | |
| Liquid |
Chemical Identifiers
| 5157-89-1 | |
| 198.306 | |
| XHXUANMFYXWVNG-DLOVCJGASA-N | |
| 21630934 | |
| CC1CCC(C(C1)OC(=O)C)C(C)C |
| C12H22O2 | |
| MFCD00062977 | |
| neoisomenthyl acetate, +-menthylacetate, 1s-2alpha-1-methylethyl-5alpha-methylcyclohexan-1alpha-ol acetate | |
| [(1S,2S,5S)-5-methyl-2-propan-2-ylcyclohexyl] acetate |
Safety and Handling
TSCA : No