missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Menthol 99.0+%, TCI America™
$32.28 - $233.52
Chemical Identifiers
| CAS | 2216-51-5 |
|---|---|
| Molecular Formula | C10H20O |
| Molecular Weight (g/mol) | 156.269 |
| MDL Number | MFCD00062979 |
| InChI Key | NOOLISFMXDJSKH-KXUCPTDWSA-N |
| Synonym | l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol |
| PubChem CID | 16666 |
| ChEBI | CHEBI:15409 |
| IUPAC Name | (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
| SMILES | CC1CCC(C(C1)O)C(C)C |
Chemical Identifiers
| 2216-51-5 | |
| 156.269 | |
| NOOLISFMXDJSKH-KXUCPTDWSA-N | |
| 16666 | |
| (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
| C10H20O | |
| MFCD00062979 | |
| l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol | |
| CHEBI:15409 | |
| CC1CCC(C(C1)O)C(C)C |
Specifications
| 2216-51-5 | |
| White | |
| C10H20O | |
| l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol | |
| CC1CCC(C(C1)O)C(C)C | |
| 156.269 | |
| CHEBI:15409 | |
| ≥99.0% (GC) | |
| (-)-Menthol |
| 43°C | |
| 216°C | |
| MFCD00062979 | |
| NOOLISFMXDJSKH-KXUCPTDWSA-N | |
| (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol | |
| 16666 | |
| 156.27 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2333
RTECSNumber : OT0700000
TSCA : Yes