missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menthol, Crystal, FCC, Spectrum™ Chemical
M1360, 2216-51-5, C10H20O
Supplier: Spectrum Chemical Mfg Cor M136012KGBL
| Quantity | 12 kg |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical Menthol, Crystal, FCC is used as a flavorant in the food industry. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 2216-51-5 | |
| 156.27 | |
| (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol |
| C10H20O | |
| NOOLISFMXDJSKH-KXUCPTDWSA-N | |
| CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O |
Specifications
| Menthol | |
| 100% | |
| Amber Glass Bottle | |
| 12 kg | |
| NOOLISFMXDJSKH-KXUCPTDWSA-N | |
| (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol | |
| FCC |
| 2216-51-5 | |
| 41-44°C | |
| C10H20O | |
| -45° to -51° | |
| CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O | |
| 156.27 | |
| Crystalline Powder or Lumps |