missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menthol, Crystal, FCC, Spectrum™ Chemical
M1360, 2216-51-5, C10H20O
$284.85 - $35,293.64
Chemical Identifiers
| CAS | 2216-51-5 |
|---|---|
| Molecular Formula | C10H20O |
| Molecular Weight (g/mol) | 156.27 |
| InChI Key | NOOLISFMXDJSKH-KXUCPTDWSA-N |
| IUPAC Name | (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol |
| SMILES | CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18604298
|
N/A | 25 g | Amber Glass Bottle |
Each for $284.85
|
|
||||
|
18604299
|
N/A | 125 g | Amber Glass Bottle |
Each for $654.30
|
|
||||
|
18604300
|
N/A | 500 g | Amber Glass Bottle |
Each for $1,811.91
|
|
||||
|
18604301
|
N/A | 12 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18604302
|
N/A | 50 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Menthol, Crystal, FCC is used as a flavorant in the food industry. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 2216-51-5 | |
| 156.27 | |
| (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol |
| C10H20O | |
| NOOLISFMXDJSKH-KXUCPTDWSA-N | |
| CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O |
Specifications
| 2216-51-5 | |
| 41-44°C | |
| -45° to -51° | |
| CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O | |
| 156.27 | |
| Crystalline Powder or Lumps |
| 100% | |
| C10H20O | |
| NOOLISFMXDJSKH-KXUCPTDWSA-N | |
| (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol | |
| FCC | |
| Menthol |