missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Navy Plaid Flame Resistant Flex Knit Button Down Shirt
Men's Navy Plaid Flame Resistant Flex Knit Button Down Shirt provides flame-resistant protection in a lightweight blended fabric.
$155.39 - $155.39
Specifications
| Gender | Men's |
|---|---|
| Material | Cotton, Spandex |
| Color | Navy |
| Neck Style | Collar |
| Sleeve Style | Long Sleeve |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-204-459
|
Bulwark™
STG2NVRGS |
Small |
Each for $155.39
|
|
|||||
|
17-204-039
|
Bulwark™
STG2NVRGM |
Medium |
Each for $155.39
|
|
|||||
|
17-204-226
|
Bulwark™
STG2NVRGL |
Large |
Each for $155.39
|
|
|||||
|
17-204-373
|
Bulwark™
STG2NVRGXL |
X-Large |
Each for $155.39
|
|
|||||
|
17-204-049
|
Bulwark™
STG2NVRGXXL |
2X-Large |
Each for $155.39
|
|
|||||
|
17-204-223
|
Bulwark™
STG2NVRG3XL |
3X-Large |
Each for $155.39
|
|
|||||
|
17-204-045
|
Bulwark™
STG2NVLNM |
Medium (Long) |
Each for $155.39
|
|
|||||
|
17-204-019
|
Bulwark™
STG2NVLNL |
Large (Long) |
Each for $155.39
|
|
|||||
|
17-204-159
|
Bulwark™
STG2NVLNXL |
X-Large (Long) |
Each for $155.39
|
|
|||||
|
17-204-355
|
Bulwark™
STG2NVLNXXL |
2X-Large (Long) |
Each for $155.39
|
|
|||||
Description
- The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
- Spandex-blend fabric allows it to stretch freely.
- Button-down collar keeps you looking crisp and sharp all day.
- Easy-to-spot CAT 2/2112 label on the front pocket and left sleeve leave no question that you're protected on the job site.
- Red buttonhole stitching and right sleeve placket leave no doubt you're armed with the very best.
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 1
- Primary Closure: Button
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
- Blend: Body: 93% Cotton / 7% Spandex Elastane
- Features: Button down collar, Single chest pocket with button closure, Lightweight, stretch fabric
- Protection: ATPV Arc Rating: 8.3
- Country of Origin: Imported, Made in USA Fabric
- Gender: Male
- Brand: Bulwark™
Specifications
| Men's | |
| Navy | |
| Long Sleeve | |
| Button | |
| 1 | |
| 8.3 |
| Cotton, Spandex | |
| Collar | |
| Knit | |
| Reusable | |
| Category 2 | |
| NFPA 2112, NFPA 70E |