missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Navy Plaid Flame Resistant Flex Knit Button Down Shirt

Men's Navy Plaid Flame Resistant Flex Knit Button Down Shirt provides flame-resistant protection in a lightweight blended fabric.

Supplier:  Bulwark™ STG2NVRG3XL

 View more versions of this product

Catalog No. 17-204-223


Only null left
Explore available promotions

Description

Description

  • The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
  • Spandex-blend fabric allows it to stretch freely.
  • Button-down collar keeps you looking crisp and sharp all day.
  • Easy-to-spot CAT 2/2112 label on the front pocket and left sleeve leave no question that you're protected on the job site.
  • Red buttonhole stitching and right sleeve placket leave no doubt you're armed with the very best.
  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 1
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
  • Blend: Body: 93% Cotton / 7% Spandex Elastane
  • Features: Button down collar, Single chest pocket with button closure, Lightweight, stretch fabric
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Male
  • Brand: Bulwark™
Specifications

Specifications

3X-Large
Cotton, Spandex
Collar
Knit
Reusable
Category 2
NFPA 2112, NFPA 70E
Men's
Navy
Long Sleeve
Button
1
8.3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions