missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Khaki Flame Resistant Flex Knit Button Down Shirt

Men's Khaki Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.

$153.18 - $153.18

Specifications

Gender Men's
Material Cotton, Spandex
Color Khaki
Neck Style Collar
Sleeve Style Long Sleeve
View More Specs

Products 10
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-204-305
View Documents
Bulwark™
STG4KHRGS
Small
Each for $153.18
Only null left
 
17-204-302
View Documents
Bulwark™
STG4KHRGM
Medium
Each for $153.18
Only null left
 
17-204-177
View Documents
Bulwark™
STG4KHRGL
Large
Each for $153.18
Only null left
 
17-204-276
View Documents
Bulwark™
STG4KHRGXL
X-Large
Each for $153.18
Only null left
 
17-203-980
View Documents
Bulwark™
STG4KHRGXXL
2X-Large
Each for $153.18
Only null left
 
17-203-975
View Documents
Bulwark™
STG4KHLNM
Medium (Long)
Each for $153.18
Only null left
 
17-204-272
View Documents
Bulwark™
STG4KHLNL
Large (Long)
Each for $153.18
Only null left
 
17-204-173
View Documents
Bulwark™
STG4KHLNXL
X-Large (Long)
Each for $153.18
Only null left
 
17-204-409
View Documents
Bulwark™
STG4KHLNXXL
2X-Large (Long)
Each for $153.18
Only null left
 
17-204-281
View Documents
Bulwark™
STG4KHRG3XL
3X-Large
Each for $153.18
Only null left
 
Description

Description

  • The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
  • Spandex-blend fabric and side seam gussets allows it to stretch freely without coming untucked.
  • Two reinforced chest pockets with button flaps allows convenient storage.
  • Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site.
  • Certifications Level 1: ASTM F1506, NFPA 70E, NFPA 2112
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 2
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
  • Body: 93% Cotton / 7% Spandex Elastane
  • Collar: Banded collar
  • Pocket: Two chest pockets with flaps and button closure
  • Features: Banded collar, Full underarm gusset for added mobility, Lightweight, stretch fabric
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Male
  • Brand: Bulwark™
Specifications

Specifications

Men's
Khaki
Long Sleeve
Button
2
8.3
Cotton, Spandex
Collar
Knit
Reusable
Category 2
ASTM F1506, NFPA 70E, NFPA 2112
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'