missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Khaki Flame Resistant Flex Knit Button Down Shirt

Catalog No. 17204305
Change view
Click to view available options
Size:
2X-Large (Long)
Large (Long)
Medium (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204305 Small
17204302 Medium
17204177 Large
17204276 X-Large
17203980 2X-Large
17203975 Medium (Long)
17204272 Large (Long)
17204173 X-Large (Long)
17204409 2X-Large (Long)
17204281 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17204305 Supplier Bulwark™ Supplier No. STG4KH RG S

Men's Khaki Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.

  • The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
  • Spandex-blend fabric and side seam gussets allows it to stretch freely without coming untucked.
  • Two reinforced chest pockets with button flaps allows convenient storage.
  • Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site.
  • Certifications Level 1: ASTM F1506, NFPA 70E, NFPA 2112
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 2
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
  • Body: 93% Cotton / 7% Spandex Elastane
  • Collar: Banded collar
  • Pocket: Two chest pockets with flaps and button closure
  • Features: Banded collar, Full underarm gusset for added mobility, Lightweight, stretch fabric
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Male
  • Brand: Bulwark™

Specifications

Size Small
Gender Men's
Material Cotton, Spandex
Color Khaki
Neck Style Collar
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance ASTM F1506, NFPA 70E, NFPA 2112
Show More Show Less