missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Khaki Flame Resistant Flex Knit Button Down Shirt
Men's Khaki Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
Supplier: Bulwark™ STG4KHRG3XL
Description
- The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
- Spandex-blend fabric and side seam gussets allows it to stretch freely without coming untucked.
- Two reinforced chest pockets with button flaps allows convenient storage.
- Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site.
- Certifications Level 1: ASTM F1506, NFPA 70E, NFPA 2112
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 2
- Primary Closure: Button
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
- Body: 93% Cotton / 7% Spandex Elastane
- Collar: Banded collar
- Pocket: Two chest pockets with flaps and button closure
- Features: Banded collar, Full underarm gusset for added mobility, Lightweight, stretch fabric
- Protection: ATPV Arc Rating: 8.3
- Country of Origin: Imported, Made in USA Fabric
- Gender: Male
- Brand: Bulwark™
Specifications
| 3X-Large | |
| Cotton, Spandex | |
| Collar | |
| Knit | |
| Reusable | |
| Category 2 | |
| ASTM F1506, NFPA 70E, NFPA 2112 |
| Men's | |
| Khaki | |
| Long Sleeve | |
| Button | |
| 2 | |
| 8.3 |