Learn More
Thermo Scientific™ Mem-PER™ Plus Membrane Protein Extraction Kit
Description
Perform small-scale solubilization and enrichment of integral membrane proteins and membrane-associated proteins in a simple reagent-based procedure using Thermo Scientific™ Mem-PER Plus Membrane Protein Extraction Kit.
Additional Information and Online Ordering
Highlights:
Extraction and isolation – produces minimal cross-contamination (typically less than 10%) of cytosolic protein into the membrane protein fraction
Cells or tissues – effective for extraction from cultured mammalian cells and mammalian tissues
Downstream compatibility – analyze membrane protein extracts by SDS-PAGE, Western blotting, immunoprecipitation and protein assays
Fast and simple – isolation of membrane proteins in approximately one hour
No special equipment required – only a benchtop microcentrifuge, tubes, homogenizers, and pipettors are needed
Includes:
Kit contains a cell wash solution, permeabilization buffer, and solubilization buffer.
Specifications
Specifications
| Product Type | Protein Extraction Kit |
| Content And Storage | Upon receipt store Cell Wash Solution and Solubilization Buffer at 4°C. Aliquot and store Permeabilization Buffer at -20°C to avoid freeze/thaws. Product is shipped on ice. |
| Form | Liquid |
| Product Line | Mem-PER |
| Reagent Type | Cell Lysis Buffer, Detergent Solution, Subcellular Fractionation |
| Sufficient For | 50 Samples |
| Quantity | 300 mL |
Frequently Asked Questions (FAQs)
The kit reagents can be scaled up or down as long as the ratio of reagents to cells is maintained. However, we don't have specific guidelines for the range of cell numbers that can be used because protein yields can vary depending on the cell line. It is important to note that the process of scaling the kit reagents up or down and determining the appropriate cell numbers for optimal protein yield will require empirical optimization by the investigator.
The composition of the Permeabilization Buffer, including the identity and concentration of any detergent therein, is proprietary. However, the Permeabilization Buffer contains a very small amount of a weak non-ionic detergent to facilitate the permeabilization of the cells and release of the cytosolic proteins.
The Permeabilization buffer in the Mem-PER Plus Membrane Protein Extraction Kit (Cat. No. 89842) creates pores in the plasma membrane of cells that releases cytosolic proteins. The permeabilized cells are centrifuged, and the supernatant contains the cytosolic protein fraction. The pellet is then resuspended in the Solubilization buffer containing a proprietary detergent that solubilizes plasma, mitochondrial, and ER membrane proteins. Subsequent centrifugation yields a supernatant containing solubilized membrane and membrane-associated proteins.
The Mem-PER Plus Membrane Protein Extraction Kit is not suitable for any organism that has a cell wall, including algae, plants, yeast, or bacteria.
M-PER Mammalian Protein Extraction Reagent can extract some membrane or cytoskeletal proteins, but the extraction efficiency is not consistent. The reagent was not intended to specifically extract these proteins. We recommend using Mem-PER Plus Membrane Protein Extraction Kit (Cat. No. 89842) for membrane protein extraction or Subcellular Protein Fractionation Kit for Cultured Cells (Cat. No. 78840) for compartmental extraction.
For Research Use Only. Not for use in diagnostic procedures.