missing translation for 'onlineSavingsMsg'
Learn More

Corning™ MEM Nonessential Amino Acid Solution

Suggested replacement for 25-025-CIR

$152.00 - $152.00

Specifications

Content And Storage 2° to 8°C
Form Liquid
Product Type Amino Acid Supplement
Sterility Sterile-filtered
pH <2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MT25025CI
View Documents
Corning™
25025CI
6 x 100 mL
Case of 6 Each for $152.00
Only null left
 
Description

Description

cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

Specifications

Specifications

2° to 8°C
Amino Acid Supplement
<2
Liquid
Sterile-filtered
Videos
SDS
Documents

Documents