missing translation for 'onlineSavingsMsg'
Learn More

Melting point Standard 283-286C, MilliporeSigma™ Supelco™

Supplier:  Merck Emd Millipore 673721G

 View more versions of this product

Catalog No. 111015673


Only null left
Explore available promotions

Description

Description

Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
Specifications

Specifications

84-65-1
7.16 (vs. air)
Neat
MFCD00001188
379°C to 381°C
Analytical Standard
208.21
1 mmHg (190°C)
185°C - closed cup
C14H8O2
283°C to 286°C (±0.3 C); 284°C to 286°C
Anthraquinone
1 g
Analytical Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions