missing translation for 'onlineSavingsMsg'
Learn More

Melting point Standard 283-286C, MilliporeSigma™ Supelco™

Catalog No. p-7187567
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015674 250 mg
111015673 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015674 Supplier Merck Emd Millipore Supplier No. 67372250MG
Only null left
Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

CAS 84-65-1
Vapor Pressure 1 mmHg (190°C)
Vapor Density 7.16 (vs. air)
Flash Point 185°C - closed cup
Physical Form Neat
Linear Formula C14H8O2
MDL Number MFCD00001188
Melting Point 283°C to 286°C (±0.3 C); 284°C to 286°C
Boiling Point 379°C to 381°C
Synonym Anthraquinone
Type Analytical Standard
Quantity 250 mg
Formula Weight 208.21
Grade Analytical Standard
Show More Show Less