missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Melanin, synthetic

Catalog No. p-7214712
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466430010 1 g
AC466430050 5 g
AC466432500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC466430010 Supplier Thermo Scientific Chemicals Supplier No. 466430010
Only null left

CAS: 8049-97-6 | C18H10N2O4, C18H10N2O4 | 318.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8049-97-6
Molecular Formula C18H10N2O4

Specifications

Quantity 1 g
Formula Weight 318.29
InChI Key XUMBMVFBXHLACL-UHFFFAOYSA-N
SMILES CC1=C2NC=C3C2=C(C2=CNC4=C(C)C(=O)C(=O)C3=C24)C(=O)C1=O
IUPAC Name 6,14-dimethyl-4,12-diazapentacyclo[8.6.1.1²,⁵.0¹³,¹⁷.0⁹,¹⁸]octadeca-1(17),2,5,9(18),10,13-hexaene-7,8,15,16-tetrone
Molecular Weight (g/mol) 318.29
Chemical Name or Material Melanin
EINECSNumber 232-473-6

RUO – Research Use Only