missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Meisoindigo

Catalog No. AC468801000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468801000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468801000 Supplier Thermo Scientific™ Supplier No. 468801000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Meisoindigo
CAS 97207-47-1
Synonym N-Methylisoindigotin; 3-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)-1,3-dihydro-1-methyl-2H-indol-2-one
Quantity 100 mg
Melting Point 230°C
Molecular Formula C17 H12 N2 O2
Formula Weight 276.29
Physical Form Crystalline Powder
Color Orange to Dark-red to Brown