missing translation for 'onlineSavingsMsg'
Learn More

Mefenamic Acid 98.0+%, TCI America™

Catalog No. M1782500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1782500G 500 g
M178225G 25 g
M1782100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1782500G Supplier TCI America Supplier No. M1782500G

Chemical Identifiers

CAS 61-68-7
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
MDL Number MFCD00051721
InChI Key HYYBABOKPJLUIN-UHFFFAOYSA-N
Synonym mefenamic acid, ponstel, ponstan, 2-2,3-dimethylphenyl amino benzoic acid, mephenamic acid, parkemed, coslan, mefacit, ponalar, methenamic acid
PubChem CID 4044
ChEBI CHEBI:6717
IUPAC Name 2-(2,3-dimethylanilino)benzoic acid
SMILES CC1=C(C(=CC=C1)NC2=CC=CC=C2C(=O)O)C

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 500 g
Formula Weight 241.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Mefenamic Acid
EINECSNumber (3)-1469
RTECSNumber CB4550000
TSCA No